Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Incoloy
®
alloy 800 - heat resisting alloy, Ni32.5Cr21Fe
Products
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF04596307
Incoloy
®
alloy 800 - heat resisting alloy, Ni32.5Cr21Fe,
500mm tube, 9.5mm outside diameter, 0.7mm wall thickness, 8.1mm inside diameter, annealed
Expand
View Pricing
GF25926699
Incoloy
®
alloy 800 - heat resisting alloy, Ni32.5Cr21Fe,
100mm tube, 9.5mm outside diameter, 0.7mm wall thickness, 8.1mm inside diameter, annealed
Expand
View Pricing
GF40836775
Incoloy
®
alloy 800 - heat resisting alloy, Ni32.5Cr21Fe,
200mm tube, 9.5mm outside diameter, 0.7mm wall thickness, 8.1mm inside diameter, annealed
Expand
View Pricing
GF02094094
Incoloy
®
alloy 800 - heat resisting alloy, Ni32.5Cr21Fe,
rod, 100mm x 10mm diameter, annealed
Expand
View Pricing
GF99325244
Incoloy
®
alloy 800 - heat resisting alloy, Ni32.5Cr21Fe,
rod, 200mm x 10mm diameter, annealed
Expand
View Pricing
GF06643166
Incoloy
®
alloy 800 - heat resisting alloy, Ni32.5Cr21Fe,
rod, 500mm x 10mm diameter, annealed
Expand
View Pricing