Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hyperfilm
™
MP
Products
Manufacturer
Length (cm)
Length (in)
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GE28-9068-46
Hyperfilm
™
MP,
W × L 8 in. × 10 in., pack of 100 ea
Expand
View Pricing
GE28-9068-44
Hyperfilm
™
MP,
W × L 18 cm × 24 cm, pack of 100 ea
Expand
View Pricing
GE28-9068-42
Hyperfilm
™
MP,
W × L 5 in. × 7 in., pack of 50 ea
Expand
View Pricing
GE28-9068-45
Hyperfilm
™
MP,
W × L 8 in. × 10 in., pack of 50 ea
Expand
View Pricing
GE28-9068-50
Hyperfilm
™
MP,
W × L 18 cm × 24 cm, pack of 50 ea (individual film envelopes)
Expand
View Pricing
GE28-9068-43
Hyperfilm
™
MP,
W × L 18 cm × 24 cm, pack of 50 ea
Expand
View Pricing