Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
HybridSPE
®
DPX Tip
Synonym(s):
HybridSPE
®
DPX SPE Tips
Products
Brand
Supelco (8)
Product Line
Chemical Composition
Volume (ml)
Volume (µL)
Application
Package Size
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
52974-U
HybridSPE
®
DPX Tip,
Tecan, Tip volume 1 mL, box of 96 tips, bed weight 50 mg
Expand
View Pricing
52982-U
HybridSPE
®
DPX Tip,
Universal format, Tip volume 1 mL, box of 96 tips, bed weight 50 mg
Expand
View Pricing
52981-U
HybridSPE
®
DPX Tip,
Universal format, Tip volume 1 mL, box of 96 tips, bed weight 30 mg
Expand
View Pricing
52979-U
HybridSPE
®
DPX Tip,
INTEGRA, Tip volume 300 μL, box of 96 tips, bed weight 30 mg
Expand
View Pricing
52978-U
HybridSPE
®
DPX Tip,
Hamilton, Tip volume 1 mL, box of 96 tips, bed weight 50 mg
Expand
View Pricing
52973-U
HybridSPE
®
DPX Tip,
Tecan, Tip volume 200 μL, box of 96 tips, bed weight 30 mg
Expand
View Pricing
52980-U
HybridSPE
®
DPX Tip,
INTEGRA, Tip volume 1.250 mL, box of 96 tips, bed weight 50 mg
Expand
View Pricing
52977-U
HybridSPE
®
DPX Tip,
Hamilton, Tip volume 300 μL, box of 96 tips, bed weight 30 mg
Expand
View Pricing