Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
HS-PEG-NH2
Synonym(s):
HS-PEG-NH2
Linear Formula:
HS(CH
2
CH
2
O)nCH
2
CH
2
NH
2
HCl
Products
Brand
Sigma-Aldrich (4)
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
JKA5143
HS-PEG-NH2,
average M
N
2,000, HCl Salt, cross-linking reagent thiol reactive, cross-linking reagent carboxyl reactive
Expand
View Pricing
JKA5145
HS-PEG-NH2,
average M
N
5,000, HCl Salt, cross-linking reagent thiol reactive, cross-linking reagent carboxyl reactive
Expand
View Pricing
JKA5144
HS-PEG-NH2,
average M
N
3,500, HCl Salt, cross-linking reagent thiol reactive, cross-linking reagent carboxyl reactive
Expand
View Pricing
JKA5146
HS-PEG-NH2,
average M
n
7,500, HCl Salt, cross-linking reagent thiol reactive, cross-linking reagent carboxyl reactive
Expand
View Pricing