Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexyl octanoate
Linear Formula:
CH
3
(CH
2
)
6
CO
2
(CH
2
)
5
CH
3
CAS Number:
1117-55-1
Molecular Weight:
228.37
Products
Product Category
aroma chemicals (2)
Brand
Sigma-Aldrich (2)
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Food Grade (2)
Kosher (2)
Fragrance grade (1)
Halal (1)
Natural (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
W257508
Hexyl octanoate,
≥97%, FG
Expand
View Pricing
W257525
Hexyl octanoate,
natural (US), ≥97%, FG
Expand
View Pricing