Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hettich
®
centrifuge buckets
Products
Manufacturer
Package Size
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z601535
Hettich
®
centrifuge buckets,
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R Centrifuges, bucket/carrier for rotors 1624; 1726
Expand
View Pricing
Z657859
Hettich
®
centrifuge buckets,
for ROTINA 38/38R and UNIVERSAL 320/320R, bucket/carrier for rotors 1494 and 1724
Expand
View Pricing
Z631167
Hettich
®
centrifuge buckets,
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R Centrifuges, bucket/carrier for rotors 1624; 1726
Expand
View Pricing
Z601470
Hettich
®
centrifuge buckets,
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R, bucket/carrier for rotor 1624; 1726
Expand
View Pricing
Z658014
Hettich
®
centrifuge buckets,
for ROTOFIX 32A and UNIVERSAL 320/320R Centrifuges, bucket/carrier for rotor 1628
Expand
View Pricing
Z601462
Hettich
®
centrifuge buckets,
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R, bucket/carrier for rotors 1624; 1726
Expand
View Pricing
Z653365
Hettich
®
centrifuge buckets,
for ROTOFIX 32A and UNIVERSAL 320/320R, bucket/carrier for rotors 1624; 1726
Expand
View Pricing