Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
GF63482193
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
100mm rod, 9.5mm diameter
Expand
View Pricing
GF93442653
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
10m wire, 0.8mm diameter, annealed
Expand
View Pricing
GF27297805
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
50m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF92879667
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
200mm rod, 9.5mm diameter
Expand
View Pricing
GF01543621
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
5m wire, 0.8mm diameter, annealed
Expand
View Pricing
GF32874059
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
20m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF20164261
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
50m wire, 0.50mm diameter, annealed
Expand
View Pricing
GF02944256
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
500mm rod, 9.5mm diameter
Expand
View Pricing
GF73885674
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
1m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF83811023
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
1m wire, 0.8mm diameter, annealed
Expand
View Pricing
GF34547283
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
20m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF66108832
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
10m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF45118043
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
5m wire, 0.50mm diameter, annealed
Expand
View Pricing
GF15449878
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
20m wire, 0.50mm diameter, annealed
Expand
View Pricing
GF27326459
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
100m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF58198191
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
10m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF73706227
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
20m wire, 0.8mm diameter, annealed
Expand
View Pricing
GF76915503
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
5m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF97503699
Hastelloy
®
X alloy, Ni49.5Cr22Fe18Mo9Co1W,
10m wire, 0.50mm diameter, annealed
Expand
View Pricing