Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hanks’ Balanced Salt solution
Products
Product Category
buffers (4)
media supplements (1)
Brand
SAFC (2)
Physical Form
Phenol Red
Quality Segment
Application
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
H6648
Hanks’ Balanced Salt solution,
Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
H8264
Hanks’ Balanced Salt solution,
Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
H9394
Hanks’ Balanced Salt solution,
Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
H9269
Hanks’ Balanced Salt solution,
With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
55037C
Hanks’ Balanced Salt solution,
HBSS Modified, with calcium, with magnesium, without phenol red, liquid, suitable for cell culture
Expand
View Pricing
55021C
Hanks’ Balanced Salt solution,
HBSS Modified, with phenol red, without calcium, without magnesium, liquid
Expand
View Pricing
H4385
Hanks’ Balanced Salt solution,
10 ×, Modified, without calcium, magnesium or sodium bicarbonate
Expand
View Pricing