Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hamilton
®
needles for LT and TLL syringes
Products
Product Category
needles (6)
Brand
Supelco (1)
Manufacturer
Length (in)
Length (mm)
Compatibility
Needle Shape
Package Size
Gauge
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z288977
Hamilton
®
needles for LT and TLL syringes,
Kel-F Hub (KF), needle size 22s ga, needle L 51 mm (2 in.), pkg of 6 ea
Expand
View Pricing
21744
Hamilton
®
needles for LT and TLL syringes,
KEL-F hub, needle size 22 ga (blunt tip), needle L 51 mm (2 in.), pkg of 6 ea
Expand
View Pricing
21741
Hamilton
®
needles for LT and TLL syringes,
Metal hub (N), needle size 22 ga (blunt tip), needle L 51 mm (2 in.), pkg of 6 ea
Expand
View Pricing
21747
Hamilton
®
needles for LT and TLL syringes,
KEL-F hub (KF), needle size 22 ga (bevel tip), needle L 51 mm (2 in.), pkg of 6 ea
Expand
View Pricing
21746
Hamilton
®
needles for LT and TLL syringes,
Metal Hub (N), needle size 22 ga (bevel tip), needle L 51 mm (2 in.), pkg of 6 ea
Expand
View Pricing
20803
Hamilton
®
needles for LT and TLL syringes,
Metal Hub (N), needle size 22 ga (cone tip, side-port), needle L 51 mm (2 in.), pkg of 6 ea
Expand
View Pricing