Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Graphene, monolayer film
Linear Formula:
C
Products
Brand
Sigma-Aldrich (9)
Color
Physical Form
Length (cm)
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
900445
Graphene, monolayer film,
1 in x 1 in on copper foil, with PMMA coating, avg. no. of layers, 1
Expand
View Pricing
798177
Graphene, monolayer film,
on TEM grid substrate (Quantifoil gold)
Expand
View Pricing
900415
Graphene, monolayer film,
10 cm x 10 cm on copper foil, avg. no. of layers, 1
Expand
View Pricing
GRTRANSFER25
Graphene, monolayer film,
Easy transfer, L × W 1 in. × 1 in.
Expand
View Pricing
900443
Graphene, monolayer film,
1 cm x 1 cm on copper foil, with PMMA coating, avg. no. of layers, 1
Expand
View Pricing
799009
Graphene, monolayer film,
1 in x 1 in on copper foil, avg. no. of layers, 1
Expand
View Pricing
773719
Graphene, monolayer film,
1 cm x 1 cm on quartz, avg. no. of layers, 1
Expand
View Pricing
773700
Graphene, monolayer film,
1 cm x 1 cm on SiO
2
/Si substrate, avg. no. of layers, 1
Expand
View Pricing
GRTRANSFER10
Graphene, monolayer film,
Easy transfer, L × W 1 cm × 1 cm
Expand
View Pricing
920118
Graphene, monolayer film,
small grain, CVD graphene on silicon wafer, 4 in diameter
Expand
View Pricing