Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs reusable CultureWell
™
gaskets
Products
Product Category
gaskets (6)
Manufacturer
Diameter (mm)
Length (mm)
Surface Area cm²
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GBL103250
Grace Bio-Labs reusable CultureWell
™
gaskets,
wells, 50, diam. × depth 3 mm × 1 mm, well capacity (3-10 μL)
Expand
View Pricing
GBL103280
Grace Bio-Labs reusable CultureWell
™
gaskets,
wells, 8, well diam. × depth 6 mm × 1 mm, well capacity (15-30 μL)
Expand
View Pricing
GBL103220
Grace Bio-Labs reusable CultureWell
™
gaskets,
wells, 2, well diam. × depth 15 mm × 1.5 mm, well capacity (300-500 μL)
Expand
View Pricing
GBL103210
Grace Bio-Labs reusable CultureWell
™
gaskets,
wells, 2, well diam. × depth 15 mm × 1 mm, well capacity (250-400 μL)
Expand
View Pricing
GBL103240
Grace Bio-Labs reusable CultureWell
™
gaskets,
wells, 4, well diam. × depth 9 mm × 1 mm, well capacity (50-100 μL)
Expand
View Pricing
GBL103230
Grace Bio-Labs reusable CultureWell
™
gaskets,
wells, 3, well L × W × depth 9.5 mm × 9.5 mm × 0.8 mm, well capacity (300-500 μL)
Expand
View Pricing