Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Grace Bio-Labs CoverWell
™
incubation chambers
Products
Manufacturer
Diameter (mm)
Volume (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GBL645501
Grace Bio-Labs CoverWell
™
incubation chambers,
chamber volume 50 μL, diam. × thickness 13 mm × 0.5 mm
Expand
View Pricing
GBL645401
Grace Bio-Labs CoverWell
™
incubation chambers,
chamber volume 20 μL, diam. × thickness 13 mm × 0.2 mm
Expand
View Pricing
GBL645402
Grace Bio-Labs CoverWell
™
incubation chambers,
chamber volume 200 μL, diam. × thickness 22 mm × 0.2 mm
Expand
View Pricing
GBL645502
Grace Bio-Labs CoverWell
™
incubation chambers,
chamber volume 500 μL, W × L × thickness 22 mm × 40 mm × 0.5 mm
Expand
View Pricing