Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GR-2 Septa
Products
Product Category
septa (6)
Brand
Supelco (6)
Diameter (mm)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
20405
GR-2 Septa,
diam. 9.5 mm ( 3/8 in.), pkg of 100 ea
Expand
View Pricing
20441
GR-2 Septa,
diam. 10 mm ( 13/32 in.)
Expand
View Pricing
20421
GR-2 Septa,
diam. 11 mm ( 7/16 in.)
Expand
View Pricing
20625
GR-2 Septa,
pkg of 250 ea
Expand
View Pricing
20413
GR-2 Septa,
diam. 12.5 mm ( 1/2 in.)
Expand
View Pricing
20627
GR-2 Septa,
pkg of 1000 ea
Expand
View Pricing