Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold Nanoshells
Synonym(s):
Gold Nanoshells
Products
Brand
Sigma-Aldrich (22)
Physical Form
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
NCXGSIR150
Gold Nanoshells,
150 nm, with Streptavidin, Bioready, OD 10, Streptavidin, in Aqueous Conjugate Buffer
Expand
View Pricing
NCXGSXR150
Gold Nanoshells,
150 nm, for Covalent Conjugation, Bioready, OD 20, Carboxyl, inaqueous 0.02 mM potassium carbonate
Expand
View Pricing
NCXGSNR15010S
Gold Nanoshells,
150 nm, for Conjugation, Bioready, Dried, NHS, Small Reaction Kit (with 10 Spin Filters), kit of 10 units
Expand
View Pricing
NCXGSGN800
Gold Nanoshells,
λ
max
, 800 nm, NanoXact, 0.05 mg/mL in water, mPEG 5 kDa (Polymer)
Expand
View Pricing
NCXGSNR1501L
Gold Nanoshells,
150 nm, for Conjugation, Bioready, Dried, NHS, Large Reaction Kit (with 1 Spin Filter), kit of 1 unit
Expand
View Pricing
NCXGSGN660
Gold Nanoshells,
λ
max
, 660 nm, NanoXact, 0.05 mg/mL in water, mPEG 5 kDa (Polymer)
Expand
View Pricing
NCXGSGH660
Gold Nanoshells,
λ
max
, 660 nm, NanoXact, 1 mg/mL in water, mPEG 5 kDa (Polymer)
Expand
View Pricing
NCXGSLH800
Gold Nanoshells,
λ
max
, 800 nm, NanoXact, 1 mg/mL in water, Carboxyl (Lipoic Acid)
Expand
View Pricing
NCXGSLN660
Gold Nanoshells,
λ
max
, 660 nm, NanoXact, 0.05 mg/mL in water, Carboxyl (Lipoic Acid)
Expand
View Pricing
NCXGSPH980
Gold Nanoshells,
λ
max
, 980 nm, NanoXact, 1 mg/mL in water, PVP 40 kDa (Polymer)
Expand
View Pricing
NCXGSLN980
Gold Nanoshells,
λ
max
, 980 nm, NanoXact, 0.05 mg/mL in water, Carboxyl (Lipoic Acid)
Expand
View Pricing
NCXGSGN980
Gold Nanoshells,
λ
max
, 980 nm, NanoXact, 0.05 mg/mL in water, mPEG 5 kDa (Polymer)
Expand
View Pricing
NCXGSLH660
Gold Nanoshells,
λ
max
, 660 nm, NanoXact, 1 mg/mL in water, Carboxyl (Lipoic Acid)
Expand
View Pricing
NCXGSGH800
Gold Nanoshells,
λ
max
, 800 nm, NanoXact, 1 mg/mL in water, mPEG 5 kDa (Polymer)
Expand
View Pricing
NCXGSPN800
Gold Nanoshells,
λ
max
, 800 nm, NanoXact, 0.05 mg/mL in water, PVP 40 kDa (Polymer)
Expand
View Pricing
NCXGSPH800
Gold Nanoshells,
λ
max
, 800 nm, NanoXact, 1 mg/mL in water, PVP 40 kDa (Polymer)
Expand
View Pricing
NCXGSLH980
Gold Nanoshells,
λ
max
, 980 nm, NanoXact, 1 mg/mL in water, Carboxyl (Lipoic Acid)
Expand
View Pricing
NCXGSPN660
Gold Nanoshells,
λ
max
, 660 nm, NanoXact, 0.05 mg/mL in water, PVP 40 kDa (Polymer)
Expand
View Pricing
NCXGSPH660
Gold Nanoshells,
λ
max
, 660 nm, NanoXact, 1 mg/mL in water, PVP 40 kDa (Polymer)
Expand
View Pricing
NCXGSGH980
Gold Nanoshells,
λ
max
, 980 nm, NanoXact, 1 mg/mL in water, mPEG 5 kDa (Polymer)
Expand
View Pricing
Page 1 of 2
Page 1 of 2