Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GLACIERbrand
™
, triple density
Products
Color
Manufacturer
Volume (ml)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z740282
GLACIERbrand
™
, triple density,
medium ice pan, volume 4 L, blue
Expand
View Pricing
Z740286
GLACIERbrand
™
, triple density,
large ice pan, volume 9 L, blue
Expand
View Pricing
Z740279
GLACIERbrand
™
, triple density,
small ice pan, volume 1 L, blue
Expand
View Pricing
Z740277
GLACIERbrand
™
, triple density,
large ice bucket, volume 5 L, blue
Expand
View Pricing
Z740274
GLACIERbrand
™
, triple density,
small ice bucket, volume 2.5 L, blue
Expand
View Pricing