Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GE Storage Phosphor Screens
Products
Manufacturer
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GE28-9564-76
GE Storage Phosphor Screens,
BAS-IP MS 3543 E, multipurpose standard, size 35 cm × 43 cm, for Typhoon FLA 9000 only
Expand
View Pricing
GE28-9564-75
GE Storage Phosphor Screens,
BAS-IP MS 2025 E, Multipurpose standard, size 20 cm × 25 cm
Expand
View Pricing
GE28-9564-74
GE Storage Phosphor Screens,
BAS-IP MS 2040 E, multipurpose standard, size 20 cm × 40 cm
Expand
View Pricing
GE28-9564-81
GE Storage Phosphor Screens,
BAS-IP TR 2040 E, tritium screen, size 20 cm × 40 cm
Expand
View Pricing
GE28-9564-77
GE Storage Phosphor Screens,
BAS-IP SR 2040 E, super resolution, size 20 cm × 40 cm
Expand
View Pricing
GE28-9564-82
GE Storage Phosphor Screens,
BAS-IP TR 2025 E, tritium screen, size 20 cm × 25 cm
Expand
View Pricing
GE28-9564-80
GE Storage Phosphor Screens,
BAS-IP SR 0813 E, super resolution, size 8 cm × 13 cm
Expand
View Pricing
GE29-0171-39
GE Storage Phosphor Screens,
BAS-IP ND 2040 E for neutron detection, size 20 cm × 25 cm
Expand
View Pricing
GE28-9564-79
GE Storage Phosphor Screens,
BAS-IP SR 12.7 x 12.7 E, super resolution, size 12.7 cm × 12.7 cm
Expand
View Pricing
GE29-0171-33
GE Storage Phosphor Screens,
BAS-IP ND 2040 E for neutron detection, size 20 cm × 40 cm
Expand
View Pricing