Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GE spacers
Products
Manufacturer
Length (cm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
GESE2619T275
GE spacers,
PVC, gray, L × thickness 10.5 mm × 0.75 mm
Expand
View Pricing
GESE2619T210
GE spacers,
PVC, white, L × thickness 10.5 mm × 1.O mm
Expand
View Pricing
GESE2119T215
GE spacers,
L × thickness 8 cm × 1.5 mm
Expand
View Pricing
GESE6118210
GE spacers,
L × W × thickness 1 cm × 16 cm × 1.0 mm
Expand
View Pricing
GESE6119215
GE spacers,
L × W × thickness 2 cm × 16 cm × 1.5 mm
Expand
View Pricing
GESE6118215
GE spacers,
L × W × thickness 1 cm × 16 cm × 1.5 mm
Expand
View Pricing
GESE2119T210
GE spacers,
L × thickness 8 cm × 1.0 mm
Expand
View Pricing
GESE2619T215
GE spacers,
PVC, gray, L × thickness 10.5 cm × 1.5 mm
Expand
View Pricing
GESE6119275
GE spacers,
L × W × thickness 2 cm × 16 cm × 0.75 mm
Expand
View Pricing
GESE6119210
GE spacers,
L × W × thickness 2 cm × 16 cm × 1.0 mm
Expand
View Pricing
GESE2119T275
GE spacers,
L × thickness 8 cm × 0.75 mm
Expand
View Pricing