Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gelatin from porcine skin
CAS Number:
9000-70-8
EC Number:
232-554-6
Products
Product Category
gelling agents (4)
proteins (2)
Biological Source
Color
pH Value
Physical Form
Tissue
Quality Segment
Application
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
G2500
Gelatin from porcine skin,
gel strength 300, Type A
Expand
View Pricing
G6144
Gelatin from porcine skin,
gel strength 80-120 g Bloom, Type A
Expand
View Pricing
G2625
Gelatin from porcine skin,
gel strength ~175 g Bloom, Type A
Expand
View Pricing
48722
Gelatin from porcine skin,
medium gel strength, suitable for microbiology
Expand
View Pricing
48724
Gelatin from porcine skin,
suitable for microbiology, high gel strength
Expand
View Pricing
48720
Gelatin from porcine skin,
suitable for microbiology, low gel strength
Expand
View Pricing
39465
Gelatin from porcine skin,
suitable for microbiology, ultrahigh gel strength
Expand
View Pricing