Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
GE glass plates
Products
Product Category
plates (7)
Manufacturer
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GESE202P10
GE glass plates,
size 10 x 8 cm
Expand
View Pricing
GESE6602
GE glass plates,
size 18 x 24 cm
Expand
View Pricing
GESE262P5
GE glass plates,
rectangular, L × D 10 cm × 10 cm
Expand
View Pricing
GESE6102
GE glass plates,
size 18 x 16 cm
Expand
View Pricing
GESE262GN5
GE glass plates,
notched, L × D 10 cm × 10 cm
Expand
View Pricing
GE80-6442-14
GE glass plates,
low fluorescence, L × D 18 cm × 16 cm, pack of 2
Expand
View Pricing
GESE6402
GE glass plates,
size 18 x 8 cm
Expand
View Pricing