Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Freezing vials, natural
Products
Product Category
vials (7)
Color
Manufacturer
Capacity (mL)
Color
Package Size
Special Grade
Natural (7)
Sterility
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
V3381
Freezing vials, natural,
capacity 500 μL, skirted bottom, sterile
Expand
View Pricing
V5381
Freezing vials, natural,
capacity 2 mL, skirted bottom, sterile
Expand
View Pricing
V4381
Freezing vials, natural,
capacity 1.5 mL, skirted bottom, sterile
Expand
View Pricing
V4131
Freezing vials, natural,
capacity 1.5 mL, conical bottom, sterile
Expand
View Pricing
V2881
Freezing vials, natural,
capacity 500 μL, skirted bottom, non-sterile
Expand
View Pricing
V4881
Freezing vials, natural,
capacity 2 mL, skirted bottom, non-sterile
Expand
View Pricing
V3881
Freezing vials, natural,
capacity 1.5 mL, skirted, non-sterile
Expand
View Pricing