
Fraxinellone

Synonym(s):
3-(3-Furyl)-3a,4,5,6-tetrahydro-3a,7-dimethylphthalide
Empirical Formula (Hill Notation):
C14H16O3
CAS Number:
Molecular Weight:
232.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing