
Fluorometholone acetate

Synonym(s):
(6α, 11bβ)-17-(Acetyloxy)-9-fluoro-11-hydroxy-6-methylpregna-1,4-diene-3,20-dione, 6α-Methyl-9α-fluoro-17-acetoxy-21-deoxyprednisolone, 6α-Methyl-9α-fluoro-21-desoxyprednisolone-17-acetate, Fluorometholone 17-acetate, NSC 47438
Empirical Formula (Hill Notation):
C24H31FO5
CAS Number:
Molecular Weight:
418.50
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2