Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fluorescein isothiocyanate isomer I
Synonym(s):
FITC, Fluorescein 5-isothiocyanate
Empirical Formula (Hill Notation):
C
21
H
11
NO
5
S
CAS Number:
3326-32-7
Molecular Weight:
389.38
EC Number:
222-042-0
Products
Product Category
fluorescence reagent (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
F7250
Fluorescein isothiocyanate isomer I,
≥90% purity (HPLC), powder
Expand
View Pricing
F4274
Fluorescein isothiocyanate isomer I,
≥97.5% purity (HPLC), powder
Expand
View Pricing