![Fluopicolide PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/161/476/8256d055-8b17-4651-97c9-2949cb1b4b8e/640/8256d055-8b17-4651-97c9-2949cb1b4b8e.png)
Fluopicolide
![Fluopicolide PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/161/476/8256d055-8b17-4651-97c9-2949cb1b4b8e/640/8256d055-8b17-4651-97c9-2949cb1b4b8e.png)
Synonym(s):
2,6-Dichloro-N-[3-chloro-5-(trifluoromethyl)-2-pyridylmethyl]benzamide
Empirical Formula (Hill Notation):
C14H8Cl3F3N2O
CAS Number:
Molecular Weight:
383.58
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2