Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Florisil
®
PR Inorganic Sorbent
Synonym(s):
Florisil
®
, Activated magnesium silicate, Magnesium silicate
Empirical Formula (Hill Notation):
MgO
3
Si
CAS Number:
1343-88-0
Molecular Weight:
100.39
EC Number:
215-681-1
Products
Product Category
resins (1)
Brand
Supelco (1)
Formula Weight
Physical Form
Separation Technique
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
46382
Florisil
®
PR Inorganic Sorbent,
PR grade (analysis acc. to FDA), 60-100 mesh
Expand
View Pricing