
Flavin adenine dinucleotide disodium salt hydrate

Synonym(s):
FAD, FAD-Na2, Riboflavin 5′-adenosine diphosphate disodium salt
Empirical Formula (Hill Notation):
C27H31N9Na2O15P2 · xH2O
CAS Number:
Molecular Weight:
829.51 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Tissue
Quality Segment
Technique
Showing 1-2 of 2