Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Flash Point Standard - Cleveland Open Cup
Products
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
certified reference material (5)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
FPCOC5
Flash Point Standard - Cleveland Open Cup,
certified reference material, ASTM D 92 (257 °C)
Expand
View Pricing
FPCOC3
Flash Point Standard - Cleveland Open Cup,
certified reference material, ASTM D 92 (148 °C)
Expand
View Pricing
FPCOC4
Flash Point Standard - Cleveland Open Cup,
certified reference material, ASTM D 92 (188 °C)
Expand
View Pricing
FPCOC6
Flash Point Standard - Cleveland Open Cup,
certified reference material, ASTM D 92 (112 °C)
Expand
View Pricing
FPCOC1
Flash Point Standard - Cleveland Open Cup,
certified reference material, ASTM D 92 (85 °C)
Expand
View Pricing