Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Finntip
®
Micro-tips 10 μL capacity
Products
Product Category
pipette tips (3)
Brand
Sigma-Aldrich (3)
Manufacturer
Length (cm)
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z369020
Finntip
®
Micro-tips 10 μL capacity,
sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing
Z369004
Finntip
®
Micro-tips 10 μL capacity,
non-sterile, pack of 1000 ea (Packed in a bag)
Expand
View Pricing
Z369012
Finntip
®
Micro-tips 10 μL capacity,
non-sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing