
Fenbendazole Related Compound A

Synonym(s):
Carbendazim, BCM, Methyl 2-benzimidazolecarbamate, Methyl benzimidazol-2-ylcarbamate
Empirical Formula (Hill Notation):
C9H9N3O2
CAS Number:
Molecular Weight:
191.19
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2