Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EZ-Pak
®
Membrane Filter
Products
Product Category
filters (10)
Brand
Millipore (10)
Agency
Diameter (mm)
Compatibility
Quality Segment
Hydrophobicity
Application
Color
Pore size (µm)
Sterility
Sterilization Method
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
EZHAWG474
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, gridded white filter, suitable for bioburden testing, input: beverage(s), bottled water
Expand
View Pricing
MSP000814
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, black filter, gridded filter, suitable for bioburden testing, input: bottled water, beverage(s)
Expand
View Pricing
EZGSWG474
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.22 μm, filter diam. 47 mm, gridded white filter, suitable for bioburden testing
Expand
View Pricing
EZAABG474
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, gridded black filter, suitable for bioburden testing, input: beverage(s)
Expand
View Pricing
EZHAWG504
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, white filter, gridded filter, General microbiological analysis
Expand
View Pricing
EZHAGG474
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, gridded green filter, suitable for bioburden testing, input: bottled water, beverage(s)
Expand
View Pricing
EZAAWG474
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, gridded white filter, suitable for bioburden testing, input: beverage(s)
Expand
View Pricing
EZHABG504
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, black filter, gridded filter, General microbiological analysis
Expand
View Pricing
EZHAGG504
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, green filter, gridded filter, General microbiological analysis
Expand
View Pricing
EZHCWG474
EZ-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.7 μm, filter diam. 47 mm, white filter, gridded filter, suitable for bioburden testing
Expand
View Pricing