Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethylbenzene
Synonym(s):
NSC 406903, Phenylethane, Ethylbenzene
Linear Formula:
C
6
H
5
C
2
H
5
CAS Number:
100-41-4
Molecular Weight:
106.17
EC Number:
202-849-4
Products
Product Category
solvents (3)
Brand
Sigma-Aldrich (3)
Supelco (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (1)
analytical standard (1)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
E12508
Ethylbenzene,
ReagentPlus
®
, 99%
Expand
View Pricing
8.01372
Ethylbenzene,
for synthesis
Expand
View Pricing
296848
Ethylbenzene,
anhydrous, 99.8%
Expand
View Pricing
1.09635
Ethylbenzene,
reference substance for gas chromatography
Expand
View Pricing
03079
Ethylbenzene,
analytical standard
Expand
View Pricing
1265457
Ethylbenzene,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR1229
Ethylbenzene,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing