
Ethyl 2-aminobenzoate

Synonym(s):
Ethyl anthranilate
Linear Formula:
H2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
165.19
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3