
Erythromycin estolate

Synonym(s):
Erythromycin 2′-propionate dodecyl sulfate
Linear Formula:
C40H71NO14 · C12H26SO4
CAS Number:
Molecular Weight:
1056.39
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade