Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Xplorer
®
Single-Channel Pipettor
Products
Product Category
pipettors (8)
Color
Manufacturer
Volume (ml)
Volume (µL)
Electronic
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
EP4861000040
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
volume 50-1000 μL, multifunction rocker switch, blue, for use with for pipette tips 1000 μL
Expand
View Pricing
EP4861000015
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
volume 0.5-10 μL, multifunction rocker switch, medium gray, for use with for pipette tips 20 mL
Expand
View Pricing
EP4861000023
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
multifunction rocker switch, yellow, volume 5-100 μL, for use with
Expand
View Pricing
EP4861000058
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
volume 0.2-5 mL, multifunction rocker switch, purple, for use with fir pipette tips 5 mL
Expand
View Pricing
EP4861000027
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
single-channel, adjustable, tip volume 10-200 μL, yellow
Expand
View Pricing
EP4861000031
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
volume 15-300 μL, multifunction rocker switch, orange, for use with for pipette tips 300 μL
Expand
View Pricing
EP4861000066
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
volume 0.5-10 mL, multifunction rocker switch, turquoise, for use with for pipette tips 10 mL
Expand
View Pricing
EP4861000017
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
single-channel, adjustable, tip volume 1-20 μL, light gray
Expand
View Pricing
EP4861000044
Eppendorf
®
Xplorer
®
Single-Channel Pipettor,
single-channel, adjustable, tip volume 0.125-2.5 mL, red
Expand
View Pricing