Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Safe-Lock micro test tubes
Products
Product Category
tubes (23)
vials (1)
Brand
Sigma-Aldrich (1)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
EP0030120086
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, colorless, pack of 1000 ea
Expand
View Pricing
EP0030120094
Eppendorf
®
Safe-Lock micro test tubes,
capacity 2.0 mL, colorless, pack of 1000 ea
Expand
View Pricing
EP0030120191
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, amber, pack of 1000 ea
Expand
View Pricing
EP0030123328
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, PCR clean, colorless, pack of 1000 ea
Expand
View Pricing
EP0030121023
Eppendorf
®
Safe-Lock micro test tubes,
capacity 0.5 mL, colorless, pack of 500 ea
Expand
View Pricing
EP0030121155
Eppendorf
®
Safe-Lock micro test tubes,
capacity 0.5 mL, amber, pack of 500 ea
Expand
View Pricing
EP0030123344
Eppendorf
®
Safe-Lock micro test tubes,
capacity 2.0 mL, PCR clean, colorless, pack of 1000 ea
Expand
View Pricing
EP0030120248
Eppendorf
®
Safe-Lock micro test tubes,
capacity 2.0 mL, amber, pack of 1000 ea
Expand
View Pricing
EP0030120175
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, blue, pack of 1000 ea
Expand
View Pricing
EP0030121708
Eppendorf
®
Safe-Lock micro test tubes,
capacity 0.5 mL, assorted colors, pack of 500 ea
Expand
View Pricing
EP0030123301
Eppendorf
®
Safe-Lock micro test tubes,
capacity 0.5 mL, PCR Clean, colorless, pack of 500 ea
Expand
View Pricing
EP0030121694
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, assorted colors, pack of 1000 ea
Expand
View Pricing
EP0030120205
Eppendorf
®
Safe-Lock micro test tubes,
tube capacity (2.0 mL), pack of 1000 ea
Expand
View Pricing
EP0030120221
Eppendorf
®
Safe-Lock micro test tubes,
capacity 2.0 mL, blue, pack of 1000 ea
Expand
View Pricing
EP0030120167
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, red, pack of 1000 ea
Expand
View Pricing
EP0030121147
Eppendorf
®
Safe-Lock micro test tubes,
capacity 0.5 mL, green, pack of 1000 ea
Expand
View Pricing
EP0030120183
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, green, pack of 1000 ea
Expand
View Pricing
EP0030121139
Eppendorf
®
Safe-Lock micro test tubes,
capacity 0.5 mL, blue, pack of 1000 ea
Expand
View Pricing
EP0030121112
Eppendorf
®
Safe-Lock micro test tubes,
capacity 0.5 mL, yellow, pack of 1000 ea
Expand
View Pricing
EP0030120159
Eppendorf
®
Safe-Lock micro test tubes,
capacity 1.5 mL, yellow, pack of 1000 ea
Expand
View Pricing
Page 1 of 2
Page 1 of 2