Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Reference 2 G
Products
Product Category
pipettors (28)
Color
Manufacturer
Volume (ml)
Volume (µL)
Package Size
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
EP4924000118
Eppendorf
®
Reference 2 G,
volume 1-10 mL, turquoise
Expand
View Pricing
EP4924000088
Eppendorf
®
Reference 2 G,
single-channel, variable, volume 100-1,000 μL, blue, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4924000100
Eppendorf
®
Reference 2 G,
volume 0.5-5 mL, purple
Expand
View Pricing
EP4924000061
Eppendorf
®
Reference 2 G,
single-channel, variable, volume 20-200 μL, yellow, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4924000096
Eppendorf
®
Reference 2 G,
single-channel, variable, volume (0.25-2.5 mL), red, includes epT.I.P.S.
®
sample bag
Expand
View Pricing
EP4924000908
Eppendorf
®
Reference 2 G,
pack of 3, Option 1, 0.5-10 uL/10-100 uL/100-1000 uL, includes [TM="epT.I.P.S." box
Expand
View Pricing
EP4925000154
Eppendorf
®
Reference 2 G,
single-channel, fixed, volume (1000 μL), blue
Expand
View Pricing
EP4924000029
Eppendorf
®
Reference 2 G,
single-channel, variable, volume 0.5-10 μL, medium gray, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4924000053
Eppendorf
®
Reference 2 G,
single-channel, variable, volume 10-100 μL, yellow, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4924000924
Eppendorf
®
Reference 2 G,
pack of 3, Option 3, 100-1000 uL/0.5-5 mL/1-10 mL, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4924000045
Eppendorf
®
Reference 2 G,
single-channel, variable, volume (2-20 μL), yellow, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4924000916
Eppendorf
®
Reference 2 G,
pack of 3, Option 2, 2-20uL/20-200 uL/100-1,000 uL, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4924000010
Eppendorf
®
Reference 2 G,
single-channel, variable, volume (0.1-2.5 μL), dark gray, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4925000162
Eppendorf
®
Reference 2 G,
single-channel, fixed, volume (2 mL), red
Expand
View Pricing
EP4925000120
Eppendorf
®
Reference 2 G,
single-channel, fixed, volume (200 μL), blue
Expand
View Pricing
EP4925000170
Eppendorf
®
Reference 2 G,
single-channel, fixed, volume (2.5 mL), red
Expand
View Pricing
EP4925000049
Eppendorf
®
Reference 2 G,
single-channel, fixed, volume (10 μL), medium gray
Expand
View Pricing
EP4924000037
Eppendorf
®
Reference 2 G,
single-channel, variable, volume (2-20 μL), light gray, includes epT.I.P.S.
®
box
Expand
View Pricing
EP4925000103
Eppendorf
®
Reference 2 G,
single-channel, fixed, volume (100 μL), yellow
Expand
View Pricing
EP4925000081
Eppendorf
®
Reference 2 G,
single-channel, fixed, volume (25 μL), yellow
Expand
View Pricing
Page 1 of 2
Page 1 of 2