Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Protein LoBind tubes
Synonym(s):
Eppendorf
®
Protein LoBind tubes
Products
Product Category
tubes (10)
Color
Manufacturer
Binding Type
Capacity (mL)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
EP0030108116
Eppendorf
®
Protein LoBind tubes,
PCR clean, withstand, 18,000 x g, capacity 1.5 mL, pack of 100 ea
Expand
View Pricing
EP0030108094
Eppendorf
®
Protein LoBind tubes,
PCR clean, withstand, 18,000 x g, capacity 0.5 mL, pack of 100 ea
Expand
View Pricing
EP0030108132
Eppendorf
®
Protein LoBind tubes,
PCR clean, withstand, 18,000 x g, capacity 2.0 mL, pack of 100 ea
Expand
View Pricing
EP0030108442
Eppendorf
®
Protein LoBind tubes,
PCR clean, withstand 18,000 x g, 1.5 mL, pack of 500 ea
Expand
View Pricing
EP0030122216
Eppendorf
®
Protein LoBind tubes,
capacity 15 mL, PCR clean, pack of 200 ea
Expand
View Pricing
EP0030108450
Eppendorf
®
Protein LoBind tubes,
PCR clean, withstand 18,000 x g, 2.0 mL, pack of 500 ea
Expand
View Pricing
EP0030122356
Eppendorf
®
Protein LoBind tubes,
capacity 5 mL, screw cap, PCR clean, pack of 200 ea (2 bags of 100 Tubes each)
Expand
View Pricing
EP0030122240
Eppendorf
®
Protein LoBind tubes,
capacity 50 mL, PCR clean, pack of 200 ea
Expand
View Pricing
EP0030122283
Eppendorf
®
Protein LoBind tubes,
capacity 25 mL, PCR clean, screw top cap, speed 17,000 x g rpm, pkg of 200 ea (4 bags x 50 tubes)
Expand
View Pricing
EP0030108531
Eppendorf
®
Protein LoBind tubes,
capacity 25 mL, PCR clean, cap (snapTec), speed 14,000 x g rpm, pkg of 200 ea (5 bags x 40 tubes)
Expand
View Pricing