Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells
Products
Product Category
plates (12)
Manufacturer
Wells
Color
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
EP0030624300
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom clear wells, PCR clean, pkg of 80 ea
Expand
View Pricing
EP0030622308
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom clear wells, sterile, pkg of 80 ea
Expand
View Pricing
EP0030627300
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom clear wells, PCR clean, pkg of 240 ea
Expand
View Pricing
EP0030621301
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
PCR clean, pkg of 80 ea, white polypropylene, V-bottom clear wells
Expand
View Pricing
EP0030623304
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom clear wells, PCR clean, pkg of 80 ea
Expand
View Pricing
EP0030621670
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
gray polypropylene, V-bottom white wells, PCR clean, pkg of 80 ea
Expand
View Pricing
EP0030625676
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
gray polypropylene, V-bottom white wells, PCR clean, pkg of 240 ea
Expand
View Pricing
EP0030625307
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom clear wells, PCR clean, pkg of 240 ea
Expand
View Pricing
EP0030626303
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
sterile, pkg of 240 ea, white polypropylene, V-bottom clear wells
Expand
View Pricing
EP0030621905
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom black wells, PCR clean, pkg of 80 ea
Expand
View Pricing
EP0030625900
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom black wells, PCR clean, pkg of 240 ea
Expand
View Pricing
EP0030628306
Eppendorf
®
Lobind Microplate 384/V-PP, 384 wells,
white polypropylene, V-bottom clear wells, PCR clean, pkg of 240 ea
Expand
View Pricing