Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
Synonym(s):
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
Products
Product Category
pipette tips (10)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
EP0030076273
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 0.1-20 μL, L 42 mm, light pink, pack of 1920 ea (5 trays of 384 tips)
Expand
View Pricing
EP0030076320
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 2-200 μL, L 53 mm, yellow, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030076290
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 0.1-10 μL, L 34 mm, dark gray, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030076346
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 50-1000 μL, L 76 mm, blue, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030076362
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 0.25–2.5 mL, L 115 mm, red, pack of 240 ea (5 trays of 48 tips)
Expand
View Pricing
EP0030076281
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 5-100 μL, L 53 mm, light yellow, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030076338
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 0-300 μL, L 55 mm, orange, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030076354
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 50-1250 μL, L 76 mm, green, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030076311
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 0.5-20 μL, L 46 mm, light gray, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030076303
Eppendorf
®
epT.I.P.S.
®
Set, Eppendorf Quality
™
,
working volume 0.1-20 μL, L 40 mm, medium gray, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing