Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
DNA LoBind tubes
Synonym(s):
Eppendorf
®
DNA LoBind tubes
Products
Product Category
tubes (10)
Color
Manufacturer
Binding Type
Capacity (mL)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
EP0030108051
Eppendorf
®
DNA LoBind tubes,
capacity 1.5 mL, PCR clean, withstand, 30,000 x g, pack of 250 ea (5 x 50EA)
Expand
View Pricing
EP0030108078
Eppendorf
®
DNA LoBind tubes,
capacity 2.0 mL, PCR clean, withstand, 25,000 x g, pack of 250 ea
Expand
View Pricing
EP0030108418
Eppendorf
®
DNA LoBind tubes,
capacity 1.5 mL, PCR clean, withstand, 30,000 x g, pack of 500 ea
Expand
View Pricing
EP0030108310
Eppendorf
®
DNA LoBind tubes,
capacity 5.0 mL, PCR clean, pack of 200 ea (4 bags of 50 ea)
Expand
View Pricing
EP0030108035
Eppendorf
®
DNA LoBind tubes,
capacity 0.5 mL, PCR clean, withstand, 30,000 x g, pack of 250 ea
Expand
View Pricing
EP0030122348
Eppendorf
®
DNA LoBind tubes,
capacity 5 mL, cap (screw cap), PCR clean, case of 200 ea (2 bags of 100 Tubes each)
Expand
View Pricing
EP0030108426
Eppendorf
®
DNA LoBind tubes,
capacity 2.0 mL, PCR clean, withstand, 30,000 x g, pack of 500 ea
Expand
View Pricing
EP0030108400
Eppendorf
®
DNA LoBind tubes,
capacity 0.5 mL, PCR clean, withstand, 30,000 x g, pack of 500 ea
Expand
View Pricing
EP0030122275
Eppendorf
®
DNA LoBind tubes,
capacity 25 mL, PCR clean, screw top cap, withstand 17,000xg, pkg of 200 ea (5 bags x 40 tubes)
Expand
View Pricing
EP0030108523
Eppendorf
®
DNA LoBind tubes,
capacity 25 mL, PCR clean, cap (snapTec), withstand 17,000xg, pkg of 200 ea (5 bags x 40 tubes)
Expand
View Pricing