Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Adapter for 500 mL Rectangular Bucket
Products
Product Category
adapters (8)
Brand
Sigma-Aldrich (1)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
EP5810739004
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
Holds 50 mL tubes, pack of 2 ea
Expand
View Pricing
EP5810722004
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
holds 12x15 mL conical tubes, pack of 2 ea
Expand
View Pricing
EP5810723000
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
holds 5 x 50 mL conical tubes, pack of 2 ea
Expand
View Pricing
EP5825722000
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
Holds 180 - 250 mL tubes, set of 2 pieces
Expand
View Pricing
EP5810747007
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
for use with 2,6-7 mL Vacutainer
Expand
View Pricing
EP5810720001
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
holds 25 x 2.6-7 mL tubes, pack of 2 ea
Expand
View Pricing
EP5810728002
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
Holds 400 mL wide neck bottles, pack of 2 ea
Expand
View Pricing
EP5810721008
Eppendorf
®
Adapter for 500 mL Rectangular Bucket,
holds 16x7-17 mL tubes, pack of 2 ea
Expand
View Pricing