Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Electrode
Products
Brand
Sigma-Aldrich (10)
Sustainability
Feature
Diameter (mm)
Surface Area cm²
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
934917
Electrode,
Thin Film Flexible Polyimide, Concentric, Outer grounded
Expand
View Pricing
934925
Electrode,
Thin Film Flexible Polyimide, Circle
Expand
View Pricing
934887
Electrode,
Flat circle 1cm
2
Expand
View Pricing
934933
Electrode,
Thin Film Flexible Polyimide, Concentric, Center grounded
Expand
View Pricing
934879
Electrode,
Flat square 1cm
2
Expand
View Pricing
937789
Electrode,
Thin Film Flexible PET, Circle 1cm
2
Expand
View Pricing
934909
Electrode,
Flat Concentric 1cm
2
Expand
View Pricing
934895
Electrode,
Flat large square 4cm
2
Expand
View Pricing
937797
Electrode,
Thin Film Flexible PET, Large Square 4cm
2
Expand
View Pricing
937800
Electrode,
Thin Film Flexible PET, Square 1cm
2
Expand
View Pricing