Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DURAN
®
graduated amber laboratory bottle
Products
Product Category
bottles (13)
Capacity (mL)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Sterility
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
DWK218066353
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (2,000 mL), non-sterile
Expand
View Pricing
DWK218064455
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (500 mL), non-sterile
Expand
View Pricing
DWK218063656
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (250 mL), non-sterile
Expand
View Pricing
DWK218065451
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (1,000 mL), non-sterile
Expand
View Pricing
DWK218062454
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (100 mL), non-sterile
Expand
View Pricing
DWK218061758
DURAN
®
graduated amber laboratory bottle,
with cap, round glass bottle, bottle capacity (50 mL), non-sterile
Expand
View Pricing
DWK218061458
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (25 mL), non-sterile
Expand
View Pricing
DWK218067358
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (5,000 mL), non-sterile
Expand
View Pricing
DWK218066953
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (3,500 mL), non-sterile
Expand
View Pricing
DWK218060856
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (10 mL), non-sterile
Expand
View Pricing
DWK218062951
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (150 mL), non-sterile
Expand
View Pricing
DWK218060807
DURAN
®
graduated amber laboratory bottle,
without cap, round amber glass bottle, bottle capacity (10 mL), non-sterile
Expand
View Pricing
DWK218065151
DURAN
®
graduated amber laboratory bottle,
with cap, round amber glass bottle, bottle capacity (750 mL), non-sterile
Expand
View Pricing