Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
Bottles, Reagent, Wide Neck
Products
Product Category
bottles (27)
Capacity (mL)
Color
Vial Bottle Color
Filter Products
Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
DWK211852403
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211885403
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211851707
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211842402
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211887301
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211857307
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211883608
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211844403
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211853605
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211881701
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211855409
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211882406
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211841706
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211884407
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211886305
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211843604
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211847306
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211854404
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
DWK211888606
Duran
®
Bottles, Reagent, Wide Neck,
amber bottle (reagent)
Expand
View Pricing
DWK211845408
Duran
®
Bottles, Reagent, Wide Neck,
clear bottle (reagent)
Expand
View Pricing
Page 1 of 2
Page 1 of 2