Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
beakers
Products
Product Category
beakers (25)
Brand
Sigma-Aldrich (6)
Manufacturer
Capacity (mL)
Height (mm)
OD (mm)
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z231843
Duran
®
beakers,
capacity 250 mL, low form
Expand
View Pricing
Z231827
Duran
®
beakers,
capacity 100 mL, low form
Expand
View Pricing
Z231878
Duran
®
beakers,
capacity 600 mL, low form
Expand
View Pricing
Z231894
Duran
®
beakers,
capacity 1,000 mL, low form
Expand
View Pricing
Z231819
Duran
®
beakers,
capacity 50 mL, low form
Expand
View Pricing
Z231835
Duran
®
beakers,
capacity 150 mL, low form
Expand
View Pricing
Z231975
Duran
®
beakers,
capacity 250 mL, Tall form
Expand
View Pricing
Z231797
Duran
®
beakers,
capacity 10 mL, low form
Expand
View Pricing
Z231800
Duran
®
beakers,
capacity 25 mL, low form
Expand
View Pricing
Z231924
Duran
®
beakers,
capacity 5,000 mL, low form
Expand
View Pricing
Z231851
Duran
®
beakers,
capacity 400 mL, low form
Expand
View Pricing
Z231908
Duran
®
beakers,
capacity 2,000 mL, low form
Expand
View Pricing
Z231789
Duran
®
beakers,
capacity 5 mL, low form
Expand
View Pricing
Z231940
Duran
®
beakers,
capacity 50 mL, Tall form
Expand
View Pricing
Z231967
Duran
®
beakers,
capacity 150 mL, Tall form
Expand
View Pricing
Z231991
Duran
®
beakers,
capacity 600 mL, Tall form
Expand
View Pricing
Z232017
Duran
®
beakers,
capacity 1,000 mL, Tall form
Expand
View Pricing
Z231959
Duran
®
beakers,
capacity 100 mL, Tall form
Expand
View Pricing
Z231916
Duran
®
beakers,
capacity 3,000 mL, low form
Expand
View Pricing
Z231886
Duran
®
beakers,
capacity 800 mL, low form
Expand
View Pricing
Page 1 of 2
Page 1 of 2