Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
baffled flask
Products
Product Category
flasks (8)
Binding Type
Capacity (mL)
Working Volume (ml)
Cap
Closure Type
Color
Package Size
Sterility
Style
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z680966
Duran
®
baffled flask,
borosilicate glass flask, working volume 250 mL, non-sterile
Expand
View Pricing
Z680974
Duran
®
baffled flask,
borosilicate glass, working volume 500 mL, non-sterile
Expand
View Pricing
DWK212833655
Duran
®
baffled flask,
Complete with membrane screw cap, neck Joints: (GL 45)
Expand
View Pricing
DWK212835459
Duran
®
baffled flask,
neck Joints: (GL 45), Complete with membrane screw cap
Expand
View Pricing
DWK212834454
Duran
®
baffled flask,
neck Joints: (GL 45), Complete with membrane screw cap
Expand
View Pricing
DWK212835401
Duran
®
baffled flask,
neck Joints: (GL 45)
Expand
View Pricing
DWK212833606
Duran
®
baffled flask,
neck Joints: (GL 45)
Expand
View Pricing
DWK212834405
Duran
®
baffled flask,
neck Joints: (GL 45)
Expand
View Pricing