Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dry-box glove/sleeve assembly
Products
Product Category
gloves (1)
Brand
Sigma-Aldrich (8)
Manufacturer
Length (in)
Color
Package Size
Special Grade
Natural (3)
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z187275
Dry-box glove/sleeve assembly,
straight sleeves, neoprene gloves, port size 8 in., size 8
Expand
View Pricing
Z187747
Dry-box glove/sleeve assembly,
straight sleeves, neoprene gloves, port size 6 in., size 9
Expand
View Pricing
Z170100
Dry-box glove/sleeve assembly,
natural rubber gloves, accordion sleeves, port size 8 in., size 10
Expand
View Pricing
Z170089
Dry-box glove/sleeve assembly,
accordion sleeves, natural rubber gloves, port size 8 in., size 8
Expand
View Pricing
Z187739
Dry-box glove/sleeve assembly,
straight sleeves, neoprene gloves, port size 8 in., size 10
Expand
View Pricing
Z187283
Dry-box glove/sleeve assembly,
straight sleeves, neoprene gloves, port size 6 in., size 8
Expand
View Pricing
Z170097
Dry-box glove/sleeve assembly,
accordion sleeves, natural rubber gloves, port size 8 in., size 9
Expand
View Pricing
Z187755
Dry-box glove/sleeve assembly,
straight sleeves, neoprene gloves, port size 6 in., size 10
Expand
View Pricing
Z187720
Dry-box glove/sleeve assembly,
straight sleeves, neoprene gloves, port size 8 in., size 9
Expand
View Pricing