Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dodecylamine
Synonym(s):
1-Aminododecane, Laurylamine, Dodecylamine
Linear Formula:
CH
3
(CH
2
)
11
NH
2
CAS Number:
124-22-1
Molecular Weight:
185.35
EC Number:
204-690-6
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
325163
Dodecylamine,
≥99%
Expand
View Pricing
D222208
Dodecylamine,
98%
Expand
View Pricing
8.03527
Dodecylamine,
for synthesis
Expand
View Pricing
44172
Dodecylamine,
puriss., ≥99.5% (GC)
Expand
View Pricing