Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DMT-dA(tac) Phosphoramidite
Synonym(s):
DMT-dA(tac) amidite
Empirical Formula (Hill Notation):
C
52
H
62
N
7
O
8
P
CAS Number:
149989-58-2
Molecular Weight:
944.06
Products
Product Category
nucleosides (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A112030
DMT-dA(tac) Phosphoramidite,
configured for ABI
Expand
View Pricing
A112000
DMT-dA(tac) Phosphoramidite,
Expand
View Pricing
A112080
DMT-dA(tac) Phosphoramidite,
configured for PerkinElmer, configured for Polygen
Expand
View Pricing