Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(±)-MDEA-D
5
solution
Empirical Formula (Hill Notation):
C
12
D
5
H
12
NO
2
CAS Number:
160227-43-0
Molecular Weight:
212.30
EC Number:
200-659-6
Products
Brand
Supelco (2)
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
certified reference material (2)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
M-068
(±)-MDEA-D
5
solution,
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing
M-067
(±)-MDEA-D
5
solution,
100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing